Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCL3L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576142 Supplier Invitrogen™ Supplier No. PA576142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene is one of several cytokine genes that are clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins that function in inflammatory and immunoregulatory processes. The protein encoded by this gene binds to several chemokine receptors, including chemokine binding protein 2 and chemokine (C-C motif) receptor 5 (CCR5). CCR5 is a co-receptor for HIV, and binding of this protein to CCR5 inhibits HIV entry. The copy number of this gene varies among individuals, where most individuals have one to six copies, and a minority of individuals have zero or more than six copies. There are conflicting reports about copy number variation of this gene and its correlation to disease susceptibility. This record represents one of two copies that are present on the ALT_REF_LOCI_2 alternate haplotype of the GRCh38 human reference genome assembly. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL3L1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CCL3L1
Gene Accession No. P16619
Gene Alias 464.2; C-C motif chemokine 3-like 1; C-C motif chemokine ligand 3 like 1; CC-chemokine (CCL3); MIP1a; CCL3L1; CCL3L3; chemokine (C-C motif) ligand 3-like 1; chemokine ligand 3-like 1; D17S1718; G0/G1 switch regulatory protein 19-2; G0S19-2; LD78; LD78BETA; LD78-beta(1-70); LD78-beta(3-70); LD78-beta(5-70); macrophage inflamatory protein 1 alpha; MIP1AP; PAT 464.2; SCYA3L; SCYA3L1; small inducible cytokine A3-like 1; Small-inducible cytokine A3-like 1; Tonsillar lymphocyte LD78 beta protein
Gene Symbols CCL3L1
Host Species Rabbit
Immunogen Recombinant fusion protein of human CCL3L1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.