Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL5 Monoclonal Antibody (OTI10G5), TrueMAB™, OriGene
SDP

Catalog No. 501681216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1216 Supplier OriGene Supplier No. TA806662
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL5 recognizes natural and recombit IP-10.

Specifications

Antigen CCL5
Applications Western Blot
Classification Monoclonal
Clone OTI10G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CCL5
Gene Accession No. P13501
Gene Alias D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
Gene Symbols CCL5
Host Species Mouse
Immunogen Full length human recombit protein of human CCL5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6352
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.