Learn More
CCL5 Mouse anti-Human, Clone: OTI2A7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806330

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CCL5 recognizes natural and recombit IP-10.Specifications
CCL5 | |
Monoclonal | |
Unconjugated | |
CCL5 | |
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP | |
Mouse | |
Affinity Chromatography | |
RUO | |
6352 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P13501 | |
CCL5 | |
Full length human recombit protein of human CCL5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.