Learn More
Invitrogen™ CCL5 (RANTES) Monoclonal Antibody (VL-1), Biotin
Mouse Monoclonal Antibody
Supplier: Invitrogen™ M230B
Description
M230B targets RANTES in ELISA, and WB applications and shows reactivity with Human samples. The M230B immunogen is recombinant human RANTES. M230B detects RANTES which has a predicted molecular weight of approximately 8 kDa. The M230B Rantes antibody (biotinylated conjugate of clone VL-1) has successfully been paired as the detection antibody in a sandwich ELISA with polyclonal capture antibody P230E. Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Biotinylated antibody M230B (clone VL-1) and antibody P230E have successfully been used in combination with recombinant Rantes protein SRNTS in ELISA applications.
CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.
Specifications
CCL5 (RANTES) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 4% BSA and proprietary preservative | |
P13501 | |
CCL5 | |
Recombinant human RANTES. | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Western Blot | |
VL-1 | |
Biotin | |
CCL5 | |
beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS d; SISd; SIS-delta; SISd; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014) | |
Mouse | |
Protein A | |
RUO | |
6352 | |
-20°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.