Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL6/C10 Antibody (262016), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB487V
Description
CCL6/C10 Monoclonal antibody specifically detects CCL6/C10 in Mouse samples. It is validated for Western Blot, Neutralization, Flow Cytometry, CyTOFSpecifications
CCL6/C10 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Neutralization, Flow Cytometry, CyTOF | |
262016 | |
Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
c10,C-C motif chemokine 6,MRP,MRP-1,Scy,Scya6 | |
E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession # P27784 | |
0.5 mL | |
Chemokines and Cytokines | |
20305 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction