Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL6/C10 Antibody (RM0183-7J13), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212420PEATT594
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Flow Cytometry.Specifications
CCL6/C10 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Rat | |
Protein A or G purified | |
Chemokines and Cytokines | |
20305.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
RM0183-7J13 | |
Flow Cytometry | |
Ccl6 | |
Mouse recombinant CCL6/C10 | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction