Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL7/MCP-3/MARC Antibody (063), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290018PCP
Description
CCL7/MCP-3/MARC Monoclonal antibody specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for ELISASpecifications
CCL7/MCP-3/MARC | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
063 | |
ELISA | |
C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
Rabbit immunized with purified, recombinant Human CCL7/MCP-3/MARC (Accession#: NP_006264.2, Gln24-Leu99) | |
0.1 mL | |
Biologically Active Proteins, Cancer, Chemokines and Cytokines | |
6354 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction