Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL8/MCP-2 Antibody (35509.11), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010391V
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CCL8/MCP-2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCL8 | |
Recombinant human monocyte chemotactic protein 2 (MCP2), expressed in E. coli. | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
6355 | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
35509.11 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
By ELISA, this antibody shows cross-reactivity with recombinant human MCP1 and recombinat mouse MIP1 alpha. In addition, it does not cross-react with other chemokines tested: rmC10, rhENA78, rhGRO alpha, rhGRO beta, rhGRO gamma, rmJE, rhI309, and rhIL8. This antibody neutralizes the biological activity of recombinant human MCP-2. Shows <20% cross-reactivity with recombinant human MCP-1 and recombinant mouse MIP-1a. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction