Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL8/MCP-2 Antibody (RM0184-17A2) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | CCL8/MCP-2 |
---|---|
Clone | RM0184-17A2 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Western Blot.Specifications
CCL8/MCP-2 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
CCL8 | |
Primary |
RM0184-17A2 | |
Western Blot | |
Unconjugated | |
Rat | |
Cytokine Research, Signal Transduction | |
Q9Z121 | |
6355 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title