Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL9 Antibody (RM0185-28B3), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212408MFV610
Description
CCL9 Monoclonal antibody specifically detects CCL9 in Mouse samples. It is validated for Western Blot, NeutralizationSpecifications
CCL9 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Neutralization | |
RM0185-28B3 | |
Western Blot, Block/Neutralize | |
CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9 | |
Mouse recombinant CCL9 | |
0.1 mL | |
Chemokines and Cytokines | |
20308 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction