Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopropylboronic Acid Mida Ester

Catalog No. 6973115G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
Catalog No. Quantity
69-731-15G 250 mg
1 options

Catalog No. 69-731-15G

Supplier: Sigma Aldrich 6973115G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclopropylboronic Acid Mida Ester

Specifications

CAS 62336-24-7
Molecular Formula C30H48BF4P2Rh
Linear Formula C30H48BF4P2Rh
Quantity 250 mg
Synonym (S,S)-Et-DUPHOS-Rh
Molecular Weight (g/mol) 660.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.