Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCM2 Antibody (OTI1E9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272235G
Description
CCM2 Monoclonal specifically detects CCM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CCM2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCM2 | |
Full length recombinant protein of human CCM2 (NP_113631) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E9 | |
Western Blot, Immunohistochemistry | |
C7orf22OSM, cerebral cavernous malformation 2, Cerebral cavernous malformations 2 protein, chromosome 7 open reading frame 22, malcavernin, MGC4067, MGC4607, MGC74868 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
83605.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction