Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCM2 Monoclonal Antibody (OTI2F9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503414S


Description
CCM2 Monoclonal antibody specifically detects CCM2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
CCM2 | |
Monoclonal | |
0.65 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BSQ5 | |
Mouse | |
Affinity chromatography | |
RUO | |
83605 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2F9 | |
Unconjugated | |
CCM2 | |
BC029157; C7orf22; Ccm2; CCM2 scaffolding protein; cerebral cavernous malformation 2; cerebral cavernous malformation 2 homolog; cerebral cavernous malformations 2 protein; Cerebral cavernous malformations protein 2 homolog; malcavernin; MGC4067; MGC4607; MGC74868; Oncostatin; OncostatinM; Osm; osmosensing scaffold for MEKK3; PP10187 | |
Full length human recombinant protein of human CCM2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction