Learn More
CCM2 Mouse anti-Human, Clone: OTI3B7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503397

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a scaffold protein that functions in the stress-activated p38 Mitogen-activated protein kinase (MAPK) signaling cascade. The protein interacts with SMAD specific E3 ubiquitin protein ligase 1 (also known as SMURF1) via a phosphotyrosine binding domain to promote RhoA degradation. The protein is required for normal cytoskeletal structure, cell-cell interactions, and lumen formation in endothelial cells. Mutations in this gene result in cerebral cavernous malformations. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CCM2 | |
Monoclonal | |
Unconjugated | |
CCM2 | |
C7orf22, OSM, PP10187 | |
Mouse | |
Affinity Chromatography | |
RUO | |
83605 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BSQ5 | |
CCM2 | |
Full length human recombit protein of human CCM2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.