Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCM2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569845 Supplier Invitrogen™ Supplier No. PA569845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

CCM2L gene ontology annotations related to this gene include cardiac atrium morphogenesis; cardiac muscle tissue growth; negative regulation of homotypic cell-cell adhesion; negative regulation of protein binding; positive regulation of fibroblast growth factor production; ventricular trabecula myocardium morphogenesis; wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCM2L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCM2l
Gene Accession No. Q9NUG4
Gene Alias C20orf160; C24H20orf160; CCM2 like scaffold protein; CCM2 like scaffolding protein; CCM2l; CCM2-like; ccml2; cerebral cavernous malformation 2; cerebral cavernous malformation 2-like; cerebral cavernous malformations 2 protein-like; dJ310O13.5; LOW QUALITY PROTEIN: cerebral cavernous malformations 2 protein-like; RGD1305202; si:dkey-223d9.2; uncharacterized protein C20orf160 homolog; cerebral cavernous malformations 2 protein-like
Gene Symbols CCM2l
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human CCM2L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140706
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.