Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCM2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552561 Supplier Invitrogen™ Supplier No. PA552561
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRRLVFPKAG RRAACRSSVS RRPLHSMPLY PPDYLIDPQI LLCDYLEKEV KFLGHLTWVT SSLNPSSRDE LLQLLDTARQ LKELPLKTTA EQDSILSLSA RCLLLTWRDN EELILRIPTH E Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

CCM2L gene ontology annotations related to this gene include cardiac atrium morphogenesis; cardiac muscle tissue growth; negative regulation of homotypic cell-cell adhesion; negative regulation of protein binding; positive regulation of fibroblast growth factor production; ventricular trabecula myocardium morphogenesis; wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCM2L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCM2l
Gene Accession No. Q9NUG4
Gene Alias C20orf160; C24H20orf160; CCM2 like scaffold protein; CCM2 like scaffolding protein; CCM2l; CCM2-like; ccml2; cerebral cavernous malformation 2; cerebral cavernous malformation 2-like; cerebral cavernous malformations 2 protein-like; dJ310O13.5; LOW QUALITY PROTEIN: cerebral cavernous malformations 2 protein-like; RGD1305202; si:dkey-223d9.2; uncharacterized protein C20orf160 homolog; cerebral cavernous malformations 2 protein-like
Gene Symbols CCM2l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCM2L. Recombinant protein control fragment (Product #RP-89709).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140706
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.