Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CCM3/PDCD10, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1016MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1016MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1016MI Supplier MilliporeSigma™ Supplier No. ABN1016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Programmed cell death protein 10 (UniProt Q9BUL8; also known as Cerebral cavernous malformations 3 protein, TF-1 cell apoptosis-related protein 15) is encoded by the PDCD10 (also known as CCM3, TFAR15) gene (Gene ID 11235) in human. PDCD10/CCM3 is an ubiquitously expressed adaptor protein that plays a role in vascularization and vessel permeability, as well as in regulating cell survival and death. Both the N-terminal dimerization domain and the C-terminal focal adhesion targeting-homology (FAT-H) domain mediate its interaction with a diverse array of signaling partners. The FAT-H domain mediates PDCD10 interaction with CCM2 to form the KRIT1 CCM2 PDCD10 complex, as well as with striatin to form the striatin interacting phosphatase and kinase (STRIPAK) complex, while the dimerization domain mediates interactions with GCKIII serine/threonine kinases (MST4/MASK, STK24/MST3 and STK25). PDCD10 overexpression leads to increased cell migration, an essential process for blood vessel formation. Loss of either PDCD10 or STK24 increases neutrophil exocytosis as a result of disrupted interaction with UNC13D, a regulator of vesicle fusion. Loss-of-function mutations in CCM1/KRIT1, CCM2/OSM, or CCM3/PDCD10 gene cause cerebral cavernous malformations (CCMs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCM3/PDCD10
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene PDCD10, CCM3, TFAR15
Gene Accession No. Q9BUL8
Host Species Rabbit
Immunogen Recombinant full-length human CCM3/PDCD10.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_009148
Test Specificity This polyclonal antibody detected the expected ∽25 kDa target band in lysate from untreated, but not CCM3 shRNA-treated, EA.hy926 human umbilical vein endothelial cells (Courtesy of Kyle M. Draheim, Ph.D., Xiaofeng Li, Ph.D., Rong Zhang, David A.Calderwood, Ph.D. and Titus J. Boggon, Ph.D., Yale University, U.S.A.).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.