Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CCMI
GREENER_CHOICESDP

Catalog No. 383750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-375-0 50 mg
38-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-375-0 Supplier R&D Systems™ Supplier No. 3837/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of α7 nAChR

Positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptors (nAChR). Evokes positive modulation of acetylcholine (ACh)-induced EC5 currents (EC50 = 0.7 μM). Exhibits cognitive-enhancing properties in rodent models; displays no cytotoxic effects in PC12 cells or rat primary cortical neurons.

Chemical Identifiers

CAS 917837-54-8
Molecular Formula C19H15Cl2N3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 388.25
Purity >98%
Molecular Weight (g/mol) 388.248
Chemical Name or Material CCMI
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.