Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCNA1 Rabbit anti-Human, Unconjugated, Clone: HOD-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19929
Description
CCNA1 Monoclonal antibody specifically detects CCNA1 in Human samples. It is validated for IP, WB-Ce, IHC-P.Specifications
CCNA1 | |
Monoclonal | |
Unconjugated | |
CCNA1 | |
A synthetic peptide corresponding to human CCNA1/CCNA2 | |
Primary | |
Human | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
HOD-3 | |
cyclin A1 | |
Rabbit | |
100 μg | |
8900, 890 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction