Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCNB1IP1 Antibody (OTI4H3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272106APC
Description
CCNB1IP1 Monoclonal specifically detects CCNB1IP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| CCNB1IP1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CCNB1IP1 | |
| Full length recombinant protein of human CCNB1IP1 (NP_878269) produced in E.coli. | |
| 0.1 mL | |
| 57820.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4H3 | |
| Flow Cytometry | |
| chromosome 14 open reading frame 18, cyclin B1 interacting protein 1, cyclin B1 interacting protein 1, E3 ubiquitin protein ligase, E3 ubiquitin-protein ligase CCNB1IP1, EC 6.3.2.-, enhancer of invasion 10, HEI10C14orf18Cyclin-B1-interacting protein 1, Human enhancer of invasion 10 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction