Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCNB1IP1 Antibody (OTI4H3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200600
Description
CCNB1IP1 Monoclonal specifically detects CCNB1IP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCNB1IP1 | |
Monoclonal | |
1.1 mg/mL | |
Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
CCNB1IP1 | |
Full length human recombinant protein of human CCNB1IP1 (NP_878269) produced in E.coli. | |
Affinity Purified | |
RUO | |
57820 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI4H3 | |
Unconjugated | |
chromosome 14 open reading frame 18, cyclin B1 interacting protein 1, cyclin B1 interacting protein 1, E3 ubiquitin protein ligase, E3 ubiquitin-protein ligase CCNB1IP1, EC 6.3.2.-, enhancer of invasion 10, HEI10C14orf18Cyclin-B1-interacting protein 1, Human enhancer of invasion 10 | |
Mouse | |
31.4 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction