Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNB1IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570394 Supplier Invitrogen™ Supplier No. PA570394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

HEI10 is a member of the E3 ubiquitin ligase family and functions in progression of the cell cycle through G(2)/M.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNB1IP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCNB1IP1
Gene Accession No. D3Z3K2
Gene Alias C14orf18; CCNB1IP1; cyclin B1 interacting protein 1; cyclin B1 interacting protein 1, E3 ubiquitin protein ligase; cyclin-B1-interacting protein 1; E3 ubiquitin-protein ligase CCNB1IP1; Gm288; HEI10; Human enhancer of invasion 10; Mei4; RING-type E3 ubiquitin transferase CCNB1IP1; Unknown (protein for MGC:134598)
Gene Symbols CCNB1IP1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse CCNB1IP1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239083
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.