Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNB1IP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101567 Supplier Invitrogen™ Supplier No. PA5101567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCNB1IP1.

HEI10 is a member of the E3 ubiquitin ligase family and functions in progression of the cell cycle through G(2)/M.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNB1IP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCNB1IP1
Gene Accession No. Q9NPC3
Gene Alias C14orf18; CCNB1IP1; cyclin B1 interacting protein 1; cyclin B1 interacting protein 1, E3 ubiquitin protein ligase; cyclin-B1-interacting protein 1; E3 ubiquitin-protein ligase CCNB1IP1; Gm288; HEI10; Human enhancer of invasion 10; Mei4; RING-type E3 ubiquitin transferase CCNB1IP1; Unknown (protein for MGC:134598)
Gene Symbols CCNB1IP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCNB1IP1(Accession Q9NPC3), corresponding to amino acid residues A201-S251.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57820
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.