Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCND1 Mouse anti-Human, Clone: DCS-6 (PE), Abnova™

Catalog No. 89034309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-309 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-309 Supplier Abnova Corporation Supplier No. MAB14208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human CCND1.

  • High specificity
  • High reactivity

Specifications

Antigen CCND1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone DCS-6
Conjugate PE
Description Mouse monoclonal antibody raised against full length recombinant human CCND1.
Dilution Flow Cytometry (5 uL/test/million cells in 0.1 mL or 5 uL/100 uL of whole blood) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P24385
Gene Alias BCL1/D11S287E/PRAD1/U21B31
Gene Symbols CCND1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CCND1.
Purification Method Protein A purification
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 595
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.