Learn More
CCND1 Mouse anti-Human, PE, Clone: SPM587, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CCND1.
Supplier: Abnova Corporation MAB13021
Description
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance throughout the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK4 or CDK6, whose activity is required for cell cycle G1/S transition. This protein has been shown to interact with tumor suppressor protein Rb and the expression of this gene is regulated positively by Rb. Mutations, amplification and overexpression of this gene, which alters cell cycle progression, are observed frequently in a variety of tumors and may contribute to tumorigenesis. [provided by RefSeq]
Specifications
CCND1 | |
Monoclonal | |
PE | |
Flow Cytometry (5 uL per test per one million cells in 0.1 mL) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user. | |
BCL1/D11S287E/PRAD1/U21B31 | |
Mouse | |
Protein A/G purification | |
RUO | |
595 | |
Store at 4°C. | |
IgG2a κ |
Flow Cytometry, Immunofluorescence | |
SPM587 | |
Mouse monoclonal antibody raised against full length recombinant human CCND1. | |
In PBS (0.05% BSA, 0.05% sodium azide). | |
CCND1 | |
Recombinant protein corresponding to full length human CCND1. | |
500 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.