Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCNDBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | CCNDBP1 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CCNDBP1 Polyclonal specifically detects CCNDBP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CCNDBP1 | |
Polyclonal | |
Rabbit | |
Human | |
23582 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SPENNDLISYNSVWVACQQMPQIPRDNKAAALLMLTKNVDFVKDAHEEMEQAVEECDPYSGLLNDTEENNSDNHNHEDDVLGFPSNQD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
cyclin D-type binding-protein 1, cyclin-D1-binding protein 1, DIP1HHM Protein, D-type cyclin-interacting protein 1, GCIPgrap2 cyclin interacting protein, Grap2 and cyclin-D-interacting protein, HHM, Human homolog of Maid, MAID protein | |
CCNDBP1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title