Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNDBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565285 Supplier Invitrogen™ Supplier No. PA565285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPENNDLISYN SVWVACQQMP QIPRDNKAAA LLMLTKNVDF VKDAHEEMEQ AVEECDPYSG LLNDTEENNS DNHNHEDDVL GFPSNQD Highest antigen sequence identity to the following orthologs - mouse 81%, rat 76%.

This gene was identified by the interaction of its gene product with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein encoded by this gene was shown to interact with cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Two alternatively spliced variants, which encode distinct isoforms, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNDBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNDBP1
Gene Accession No. O95273
Gene Alias AU022347; Ccndbp1; cyclin D1 binding protein 1; cyclin D-type binding-protein 1; cyclin-D1-binding protein 1; DIP1; D-type cyclin-interacting protein 1; GCIP; grap2 and cyclin D interacting protein; grap2 and cyclin-D-interacting protein; HHM; human homolog of Maid; Maid; Maternal Id-like protein; maternal inhibition of differentiation; SECC-8; SSEC-8; Stage specific embryonic cDNA-8 protein
Gene Symbols CCNDBP1
Host Species Rabbit
Immunogen Recombinant Human CCNDBP1. Recombinant protein control fragment (Product #RP-104728).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.