Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNDBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142024
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142024 Supplier Invitrogen™ Supplier No. PA5142024
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise both human isoforms of this protein. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

This gene was identified by the interaction of its gene product with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein encoded by this gene was shown to interact with cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Two alternatively spliced variants, which encode distinct isoforms, have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNDBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CCNDBP1
Gene Accession No. O95273
Gene Alias AU022347; Ccndbp1; cyclin D1 binding protein 1; cyclin D-type binding-protein 1; cyclin-D1-binding protein 1; DIP1; D-type cyclin-interacting protein 1; GCIP; grap2 and cyclin D interacting protein; grap2 and cyclin-D-interacting protein; HHM; human homolog of Maid; Maid; Maternal Id-like protein; maternal inhibition of differentiation; SECC-8; SSEC-8; Stage specific embryonic cDNA-8 protein
Gene Symbols CCNDBP1
Host Species Goat
Immunogen Peptide with sequence C-NRIKELTQSELEL (aa 348-360).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23582
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.