Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566140 Supplier Invitrogen™ Supplier No. PA566140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSEQPSCQQIV STTHTSSYTL QTCPAGFQTS VQGLGHMQTG VGMSLAIPVE VKPCLSVSYN RSYQINEHYP CITP Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

CCNJ is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNJ
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNJ
Gene Accession No. Q5T5M9
Gene Alias bA690P14.1; CCNJ; cyclin J; cyclin-J; Cyclin-J-like protein; D430039C20Rik
Gene Symbols CCNJ
Host Species Rabbit
Immunogen Recombinant Human CCNJ. Recombinant protein control fragment (Product #RP-106823).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54619
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.