Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCNJL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562107
Description
Immunogen sequence: TTLAQFQTPV QDLCLAYRDS LQAHRSGSLL SGSTGSSLHT PYQPLQPLDM CPVPVPASLS MHMAIAAEPR HCLATTYGSS Y Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
CCNJL is a protein coding gene and gene ontology (GO) annotation includes cyclin-dependent protein kinase holoenzyme complex; cytoplasm; nucleus; cyclin-dependent protein serine/threonine kinase regulator activity; protein kinase binding; mitotic cell cycle phase transition; protein phosphorylation; regulation of cyclin-dependent protein serine/threonine kinase activity.
Specifications
CCNJL | |
Polyclonal | |
Unconjugated | |
CCNJL | |
Ccnjl; cyclin J like; cyclin J-like; cyclin-J-like protein; Gm877 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IV13 | |
CCNJL | |
Recombinant protein corresponding to Human CCNJL. Recombinant protein control fragment (Product #RP-101286). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction