Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCNK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581885
Description
Immunogen sequence: DHTKPCWYWD KKDLAHTPSQ LEGLDPATEA RYRREGARFI FDVGTRLGLH YDTLATGIIY FHRFYMFHSF KQFPRYVTGA CCLFLAGKVE ETPKKCKDII KTARSLLNDV QFGQFGDDPK EEVMVLERIL LQTIKFDLQV EHPYQFLL.
Regulatory subunit of cyclin-dependent kinases that mediates activation of target kinases. Plays a role in transcriptional regulation via its role in regulating the phosphorylation of the C-terminal domain (CTD) of the large subunit of RNA polymerase II (POLR2A).
Specifications
CCNK | |
Polyclonal | |
Unconjugated | |
CCNK | |
AW123198; AW413594; CCNK; CPR4; CycK; cyclin K; cyclin-K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8812 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75909 | |
CCNK | |
Recombinant protein corresponding to Human CCNK. Recombinant protein control fragment (Product #RP-89012). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction