Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCNK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114710
Description
Antibody detects endogenous levels of total CCNK.
Regulatory subunit of cyclin-dependent kinases that mediates activation of target kinases. Plays a role in transcriptional regulation via its role in regulating the phosphorylation of the C-terminal domain (CTD) of the large subunit of RNA polymerase II (POLR2A).
Specifications
| CCNK | |
| Polyclonal | |
| Unconjugated | |
| CCNK | |
| AW123198; AW413594; CCNK; CPR4; CycK; cyclin K; cyclin-K | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12454, 8812 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O75909, O88874 | |
| CCNK | |
| A synthesized peptide derived from human CCNK(Accession O75909), corresponding to amino acid residues M1-L38. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction