Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNK Polyclonal Antibody, Invitrogen™

Catalog No. PIPA585020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585020 Supplier Thermo Scientific Supplier No. PA585020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSYMTGMST TSSYMSGEGY QSLQSMMKTE GPSYGALPPA YGPPAHLPYH PHVYPPNP

CCNK Polyclonal antibody specifically detects CCNK in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNK
Gene Accession No. O75909
Gene Alias AW123198; AW413594; CCNK; CPR4; CycK; cyclin K; cyclin-K
Gene Symbols CCNK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCNK
Purification Method Anitgen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.