Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104462 Supplier Invitrogen™ Supplier No. PA5104462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cyclin L1.

FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintenance of healthy cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ccnl1
Gene Accession No. Q52KE7, Q9R1Q2, Q9UK58
Gene Alias 2610030E23Rik; ANIA6A; ania-6a; AU018493; BM-001; Ccn1; Ccnl; CCNL1; Cyclin Ania-6a; cyclin L ania-6a; cyclin L gamma; cyclin L1; cyclin-L; Cyclin-L1; LOW QUALITY PROTEIN: cyclin-L1; PRO1073; UNQ530/PRO1073
Gene Symbols Ccnl1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCNL1(Accession Q9UK58), corresponding to amino acid residues R471-H521.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114121, 56706, 57018
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.