Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNY Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen CCNY
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1580720
SDP
View Documents
Novus Biologicals
NBP15807120UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP158071
SDP
View Documents
Novus Biologicals
NBP158071
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

CCNY Polyclonal specifically detects CCNY in Human samples. It is validated for Western Blot.
Specifications

Specifications

CCNY
Polyclonal
Rabbit
Cell Cycle and Replication
CBCP1C10orf9cyc-Y, CCNX, CFP 1, CFP1FLJ95513, chromosome 10 open reading frame 9, Cyclin box protein 1, Cyclin fold protein 1, cyclin Y, cyclin-box carrying protein 1, cyclin-X, cyclin-Y, Cyc-Y
CCNY
IgG
Western Blot
Unconjugated
RUO
Q8ND76
219771
Synthetic peptides corresponding to CCNY(cyclin Y) The peptide sequence was selected from the middle region of CCNY. Peptide sequence DENLHPLSKSEVPPDYDKHNPEQKQIYRFVRTLFSAAQLTAECAIVTLVY.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.