Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCNY Antibody (OTI12F10), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272462AF405
Description
CCNY Monoclonal specifically detects CCNY in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
CCNY | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCNY | |
Full length recombinant protein of human CCNY (NP_859049) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI12F10 | |
Western Blot | |
CBCP1C10orf9cyc-Y, CCNX, CFP 1, CFP1FLJ95513, chromosome 10 open reading frame 9, Cyclin box protein 1, Cyclin fold protein 1, cyclin Y, cyclin-box carrying protein 1, cyclin-X, cyclin-Y, Cyc-Y | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
219771.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction