Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCP Rabbit anti-Chemical, Polyclonal, Bioss
SDP

Catalog No. 501980925
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0925 Supplier Bioss Supplier No. BS1053R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Citrulline is naturally produced in the body as part of the metabolism of the amino acid arginine. However, in joints with rheumatoid arthritis (RA), this conversion may occur at a higher rate. Cyclic citrullinated peptide antibodies are autoantibodies produced by the immune system that are directed against cyclic citrullinated peptides (CCP). This test detects and measures anti-CCP antibodies in the blood.

Specifications

Antigen CCP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide of CCP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.