Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101547 Supplier Invitrogen™ Supplier No. PA5101547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CBCP2.

Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AGBL2
Gene Accession No. Q5U5Z8, Q8CDK2
Gene Alias 4930524K04; A430081C19Rik; Agbl2; ATP/GTP binding protein like 2; ATP/GTP binding protein-like 2; ATP/GTP-binding protein-like 2; carboxypeptidase 2, cytosolic; CBCP2; CCP2; cytoplasmic carboxypeptidase 2; cytosolic carboxypeptidase 2; testis tissue sperm-binding protein Li 96mP; testis-expressed protein 25; zebrafish testis-expressed 25; zgc:165648; zte25
Gene Symbols AGBL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human AGBL2(Accession Q5U5Z8), corresponding to amino acid residues L789-K839.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 271813, 79841
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.