Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550311 Supplier Invitrogen™ Supplier No. PA550311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CCP3 / AGBL3 protein.

Metallocarboxypeptidase that mediates both deglutamylation and deaspartylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates or polyaspartates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Agbl3
Gene Accession No. Q8CDP0, Q8NEM8
Gene Alias 2900053G10Rik; 4930431N21Rik; 6530406M24Rik; Agbl3; ATP/GTP binding protein like 3; ATP/GTP binding protein-like 3; ATP/GTP-binding protein-like 3; CBCP3; CCP3; Cytosolic carboxypeptidase 3; RGD1561995
Gene Symbols Agbl3
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human ATP/GTP binding protein-like 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340351, 76223
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.