Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550543 Supplier Invitrogen™ Supplier No. PA550543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total AGBL1 protein.

Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates from the carboxy-terminus of target proteins such as MYLK. Acts as a long-chain deglutamylase and specifically shortens long polyglutamate chains, while it is not able to remove the branching point glutamate, a process catalyzed by AGBL5/CCP5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene AGBL1
Gene Accession No. Q09M05, Q96MI9
Gene Alias AGBL1; ATP/GTP binding protein like 1; ATP/GTP binding protein-like 1; ATP/GTP-binding protein-like 1; carboxypeptidase 4, cytosolic; Ccp4; Cytosolic carboxypeptidase 4; D430020F16; EG244071; FECD8; Nna1-l1
Gene Symbols AGBL1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human ATP/GTP binding protein-like 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123624, 244071
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.