Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCPG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144677 Supplier Invitrogen™ Supplier No. PA5144677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HCT116 cell lysate, A549 cell lysate, Hela cell lysate, THP-1 cell lysate, SW480 cell lysate, HEK-293 cell lysate, SW480. Predicted band size: 87 kDa Subcellular Location: Cytoplasmic granule membrane.

Acts as an assembly platform for Rho protein signaling complexes. Limits guanine nucleotide exchange activity of MCF2L toward RHOA, which results in an inhibition of both its transcriptional activation ability and its transforming activity. Does not inhibit activity of MCF2L toward CDC42, or activity of MCF2 toward either RHOA or CDC42. May be involved in cell cycle regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCPG1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CCPG1
Gene Accession No. Q9ULG6
Gene Alias 1700030B06Rik; 1810073J13Rik; 9430028F23Rik; AI426686; AI875170; CCP8; Ccpg1; cell cycle progression 1; cell cycle progression 8 protein; cell cycle progression protein 1; Cell cycle progression restoration protein 8; CPR8; D9Ertd392e; KIAA1254; Unknown (protein for IMAGE:8060449)
Gene Symbols CCPG1
Host Species Rabbit
Immunogen Recombinant protein within human CCPG1 aa 241-390/757.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.