Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100719
Description
CCR1 Polyclonal specifically detects CCR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CCR1 | |
Polyclonal | |
Unconjugated | |
10 mM KHPO4, 140 mM NaCl, BSA with 0.1% Sodium Azide | |
CCR1 | |
Synthetic peptide: TEDYDTTTEFDYGDATPC, corresponding to amino acids 7 - 24 of Human CCR1. | |
0.05 mg | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1230 | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
1.0 mg/mL | |
Western Blot 1:1000, ELISA 1:100000, Immunohistochemistry 1:200 | |
C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Peptide sequence is < 50 % identical to other chemokine receptors in this region. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction