Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
$172.84 - $474.44
Specifications
Antigen | CCR1 |
---|---|
Clone | 984303 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1451SP
|
R&D Systems
MAB1451SP |
25 μg |
Each for $172.84
|
|
|||||
MAB1451100
|
R&D Systems
MAB1451100 |
100 μg |
Each for $474.44
|
|
|||||
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Flow Cytometry.Specifications
CCR1 | |
Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Supernatant | |
Human | |
P32246 | |
1230 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
984303 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
Mouse myeloma cell line NS0-derived recombinant human CCR1 Met1-Phe355 Accession # P32246 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CCR1 in direct ELISAs. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title