Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR1 Rabbit anti-Human, Clone: 3, Novus Biologicals™

Rabbit Monoclonal Antibody
$366.00 - $572.50
Specifications
Antigen | CCR1 |
---|---|
Clone | 3 |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR1 | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
A synthetic peptide corresponding to the center region of the Human CCR1 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
0.2 um filtered solution in PBS | |
1230 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title