Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCR1 Recombinant Rabbit Monoclonal Antibody (002)
GREENER_CHOICE

Catalog No. PIMA535975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535975 Supplier Invitrogen™ Supplier No. MA535975
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

CCR1 Recombinant Monoclonal Antibody for Western Blot, IP

Chemokines play important roles in inflammation and are critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines. One of the CCR receptors, CCR1, is expressed on neutrophils, monocytes, lymphocytes, and eosinophils and binds the leukocyte chemoattractant and hemopoiesis regulator macrophage-inflammatory protein (MIP-1 ), eotaxin, as well as several other related chemokines. Mice lacking the chemokine receptor CCR1 have defects in neutrophil trafficking and proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR1
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 002
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCR1
Gene Accession No. P32246
Gene Alias C-C chemokine receptor type 1; C-C CKR-1; C-C motif chemokine receptor 1; CC-CKR-1; Ccr1; CCR-1; CD191; chemokine (C-C motif) receptor 1; chemokine (C-C) receptor 1; CKR1; CKR-1; CMKBR1; CMKR1; HM145; LD78 receptor; macrophage inflammatory protein 1-alpha receptor; macrophage inflammatory protein-1 alpha receptor; MIP-1 alpha R; MIP-1 alphaR; MIP-1alpha-R; MIP1aR; Mip-1a-R; RANTES receptor; RANTES-R; SCYAR1
Gene Symbols CCR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human CCR1/CD191.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1230
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.