Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCR11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533409
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533409 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533409 Supplier Invitrogen™ Supplier No. PA533409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Rat, Bovine (84%).

Receptors for the C-C chemokine family include CCR-1, CCR-2A, CCR-3, CCR-4, CCR-5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. CCR11 in particular is expressed in the heart, lung, and small intestine. It binds members of the monocyte chemoattractant family including MCP-1, MCP-2, and MCP-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ACKR4
Gene Accession No. Q9NPB9
Gene Alias A630091E18Rik; Ackr4; Atypical chemokine receptor 4; C-C chemokine receptor type 11; CC chemokine receptor-like 1; C-C CKR-11; CCBP2; CC-CKR-11; CCR10; CCR11; CCR-11; Ccrl1; CCX CKR; CCX-CKR; CCX-CKR1; chemocentryx chemokine receptor; chemokine (C-C motif) receptor-like 1; chemokine (C-C) receptor-like 1; chemokine receptor CCR11; chemokine, cc motif, receptor-like protein 1; CKR-11; Cmkbrl1; orphan seven-transmembrane receptor, chemokine related; PPR1; VSHK1
Gene Symbols ACKR4
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 2nd cytoplasmic domain of human CCRL1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51554
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.