Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCR11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106552 Supplier Invitrogen™ Supplier No. PA5106552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCRL1.

Receptors for the C-C chemokine family include CCR-1, CCR-2A, CCR-3, CCR-4, CCR-5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. CCR11 in particular is expressed in the heart, lung, and small intestine. It binds members of the monocyte chemoattractant family including MCP-1, MCP-2, and MCP-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACKR4
Gene Accession No. Q924I3, Q9NPB9
Gene Alias A630091E18Rik; Ackr4; Atypical chemokine receptor 4; C-C chemokine receptor type 11; CC chemokine receptor-like 1; C-C CKR-11; CCBP2; CC-CKR-11; CCR10; CCR11; CCR-11; Ccrl1; CCX CKR; CCX-CKR; CCX-CKR1; chemocentryx chemokine receptor; chemokine (C-C motif) receptor-like 1; chemokine (C-C) receptor-like 1; chemokine receptor CCR11; chemokine, cc motif, receptor-like protein 1; CKR-11; Cmkbrl1; orphan seven-transmembrane receptor, chemokine related; PPR1; VSHK1
Gene Symbols ACKR4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACKR4(Accession Q9NPB9), corresponding to amino acid residues P300-I350.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252837, 51554, 685243
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.