Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR2 Antibody (2D8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.72 - $492.89
Specifications
| Antigen | CCR2 |
|---|---|
| Clone | 2D8 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP235333SS
![]() |
Novus Biologicals
NBP235333SS |
0.025 mg |
Each for $181.72
|
|
|||||
NBP235333
![]() |
Novus Biologicals
NBP235333 |
0.1 mg |
Each for $492.89
|
|
|||||
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CCR2 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| P41597 | |
| 729230 | |
| A synthetic peptide made to an N-terminal portion of the human CCR2 protein (between residues 1-100) [Uniprot: P41597] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2D8 | |
| Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| CCR2 | |
| IgG2b λ | |
| Protein G purified | |
| Immunogen's sequence similarity with other species: Mouse (57%), Rabbit (57%). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title