Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR2 Antibody (3G7) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen CCR2
Clone 3G7
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34274535), Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP235334SS
SDP
View Documents
Novus Biologicals
NBP235334SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP235334
SDP
View Documents
Novus Biologicals
NBP235334
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

CCR2 Monoclonal specifically detects CCR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CCR2
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
P41597
729230
A synthetic peptide made to an N-terminal portion of the human CD63 protein (between residues 1-100) [UniProt P41597]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
3G7
Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34274535), Immunohistochemistry-Paraffin 5 ug/ml
Monoclonal
Purified
RUO
Human, Mouse
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
CCR2
IgG2b λ
Protein G purified
Immunogen's sequence similarity with other species: Mouse (57%), Rabbit (57%).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.