Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP148338AF750
Description
CCR2 Polyclonal specifically detects CCR2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
CCR2 | |
Polyclonal | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
Rabbit | |
0.1 mL | |
729230 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CCR2 | |
Synthetic peptide made to an N-terminal portion of the mouse CCR2 protein (within residues 20-100). [Swiss-ProtNo. P51683] | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction